Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Phenylbenzamidine, 97%

Catalog No. aaa1321906
Click to view available options
Quantity:
5 g
25 g

Catalog No. AAA1321906

Supplier: Thermo Scientific Chemicals A1321906

Only null left
Add to Cart

CAS: 1527-91-9 | C13H12N2 | 196.253 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1527-91-9
Molecular Formula C13H12N2
Molecular Weight (g/mol) 196.253
MDL Number MFCD00019730
InChI Key MPYOKHFSBKUKPQ-UHFFFAOYSA-N
Synonym n-phenylbenzamidine, n-phenylbenzenecarboximidamide, n'-phenylbenzamidine, n1-phenylbenzene-1-carboximidamide, n2-phenylbenzamidine, n-phenylbenzimidamide, acmc-20ak1f, maybridge1_004766, iminophenylmethyl phenylamine, n-phenyl-benzenecarboximidamide
PubChem CID 200127
IUPAC Name N'-phenylbenzenecarboximidamide
SMILES C1=CC=C(C=C1)C(=NC2=CC=CC=C2)N

Specifications

Melting Point 117°C to 120°C
Quantity 5 g
Beilstein 608471
Sensitivity Hygroscopic
Formula Weight 196.25
Percent Purity 97%
Chemical Name or Material N-Phenylbenzamidine
Hazard Category H302+H312+H332-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.