Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Phenylcarbazole, 97%

Catalog No. p-4813588
Click to view available options
Quantity:
5 g

CAS: 1150-62-5 | C18H13N | 243.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1150-62-5
Molecular Formula C18H13N
Molecular Weight (g/mol) 243.31
MDL Number MFCD00004965
InChI Key VIJYEGDOKCKUOL-UHFFFAOYSA-N
PubChem CID 70851
IUPAC Name 9-phenylcarbazole
SMILES C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Melting Point 94.0°C to 97.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 5 g
Beilstein 20,I,164
Synonym n-phenylcarbazole, 9-phenyl-9h-carbazole, 9h-carbazole, 9-phenyl, carbazole, 9-phenyl, carbazol, 9-phenyl, 9-phenyl carbazole, pubchem23849, 9h-carbazole,9-phenyl, 9-phenylcarbazole, acmc-2099pd
Formula Weight 243.31
Percent Purity 97%
Chemical Name or Material N-Phenylcarbazole
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTRE or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee
EINECSNumber 214-564-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.