Learn More
n-Tetracontane, 97% min
Description
n-Tetracontane is used in a passivation layer in copper-phthalocyanine field-effect transistors for improving the charge carrier mobility and used to modify the quasi two-dimensional surface electron system on Au(111).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 80°C to 84°C |
| Color | White |
| Density | 0.779 g/mL |
| Boiling Point | 150°C (0.001 mmHg) |
| Linear Formula | CH3(CH2)38CH3 |
| Quantity | 2 g |
| Beilstein | 1800152 |
| Solubility Information | Soluble in chloroform. Slightly soluble in methanol.Insoluble in water. |
| Formula Weight | 563.09 |
| Percent Purity | ≤97% |
| Show More |
Safety and Handling
| EINECSNumber | 224-055-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.