Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Tetradecanamide, 98%

Catalog No. p-7026556
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 638-58-4 | C14H29NO | 227.392 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 638-58-4
Molecular Formula C14H29NO
Molecular Weight (g/mol) 227.392
MDL Number MFCD00025533
InChI Key QEALYLRSRQDCRA-UHFFFAOYSA-N
Synonym myristamide, n-tetradecanamide, myristic amide, myristic acid amide, tetradecylamide, unii-a269j8qg0o, myristicamide, tetradecanoic acid amide, chembl88158
PubChem CID 69492
IUPAC Name tetradecanamide
SMILES CCCCCCCCCCCCCC(=O)N

Specifications

Melting Point 105°C to 107°C
Quantity 5 g
Formula Weight 227.39
Percent Purity 98%
Chemical Name or Material n-Tetradecanamide
EINECSNumber 211-343-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.