Learn More
n-Tetradecane, 99%
Description
n-Tetradecane is used to study the thermodiffusion behavior of the microemulsion droplets of water, n-alkanes and pentaethylene glycol monododecyl ether. It is used as reference material in gas chromatographic analysis, as a solvent and finds applications in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
n-Tetradecane is used to study the thermodiffusion behavior of the microemulsion droplets of water, n-alkanes and pentaethylene glycol monododecyl ether. It is used as reference material in gas chromatographic analysis, as a solvent and finds applications in organic synthesis.
Solubility
Soluble in ether, acetone and alcohol. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | 6°C |
Density | 0.763 |
Boiling Point | 252°C to 254°C |
Flash Point | 99°C (211°F) |
Odor | Mild |
Assay Percent Range | 99% |
Refractive Index | 1.429 |
Linear Formula | CH3(CH2)12CH3 |
Quantity | 2500 g |
Solubility Information | Soluble in ether,acetone and alcohol. Insoluble in water. |
Show More |
Safety and Handling
Hazard Category | H304 |
Hazard Statement | GHS H Statement H304-H336 May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. |
Precautionary Statement | P301+P310-P331-P501c |
EINECSNumber | 211-096-0 |
RTECSNumber | XB8000000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.