Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Undecane, 99%

Catalog No. p-7049945
Click to view available options
Quantity:
50 g
250 g

CAS: 1120-21-4 | C11H24 | 156.31 g/mol

n-Undecane is used as a mild sex attractant for various types of moths and cockroaches. It is also used as an internal standard in the gas chromatographic analysis. Further, it is used as lubricants and lubricant additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-21-4
Molecular Formula C11H24
Molecular Weight (g/mol) 156.31
MDL Number MFCD00008959
InChI Key RSJKGSCJYJTIGS-UHFFFAOYSA-N
Synonym n-undecane, hendecane, n-hendecane, decane, methyl, unii-jv0qt00nue, ccris 3796, jv0qt00nue, undecane, undecane, analytical standard, und
PubChem CID 14257
ChEBI CHEBI:46342
IUPAC Name undecane
SMILES CCCCCCCCCCC

Specifications

Melting Point -26°C
Color Colorless
Density 0.74 g/mL
Boiling Point 196°C
Flash Point 60°C (140°F)
Odor Mild
Refractive Index 1.4173
Linear Formula CH3(CH2)9CH3
Quantity 50 g
UN Number UN2330
Solubility Information Miscible with ethyl alcohol and ether. Immiscible with water.
Formula Weight 156.3
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material n-Undecane
Show More Show Less
Hazard Category H227-H304
Hazard Statement GHS H Statement
H304-H226-H315-H319-H335
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P280-P301+P310-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: UNDECANE
EINECSNumber 214-300-6
RTECSNumber YQ1525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.