Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-WASP, p Ser484, p Ser485 Antibody, Novus Biologicals™
SDP

Catalog No. NB019420 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019420 0.025 mL
NBP229669 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019420 Supplier Novus Biologicals Supplier No. NBP2296690.025ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

N-WASP Polyclonal specifically detects N-WASP in Human, Rat samples. It is validated for Western Blot, ELISA.

Specifications

Antigen N-WASP (p Ser484, p Ser485)
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:2000
Formulation 100 ul phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA. with 0.05% Sodium Azide
Gene Accession No. O00401
Gene Alias N-WASPneural WiskNWASP, WiskDKFZp779G0847, WiskMGC48327
Gene Symbols WASL
Host Species Rabbit
Immunogen Phospho-N-WASP (S484/S485) synthetic peptide (coupled to KLH) corresponding to amino acid residues around serine 484 and 485 of human N-WASP. The human WASP sequence has a similar peptide sequence surrounding serine 483 and 484.
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 8976
Test Specificity The antibody detects a 63 kDa protein corresponding to the molecular weight of phosphorylated WASP on SDS-PAGE immunoBlots of Jurkat cell lysate. In rat brain, A431, human endothelial and SKN-SH cells, this antibody detects a 65 kDa protein corresponding to phosphorylated N-WASP.
Target Species Human, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.