Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N-WASP, p Ser484, p Ser485 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$243.50 - $670.50
Specifications
Antigen | N-WASP (p Ser484, p Ser485) |
---|---|
Dilution | Western Blot 1:1000, ELISA 1:2000 |
Applications | ELISA, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
N-WASP Polyclonal specifically detects N-WASP in Human, Rat samples. It is validated for Western Blot, ELISA.Specifications
N-WASP (p Ser484, p Ser485) | |
ELISA, Western Blot | |
Unconjugated | |
RUO | |
Human, Rat | |
O00401 | |
8976 | |
Phospho-N-WASP (S484/S485) synthetic peptide (coupled to KLH) corresponding to amino acid residues around serine 484 and 485 of human N-WASP. The human WASP sequence has a similar peptide sequence surrounding serine 483 and 484. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot 1:1000, ELISA 1:2000 | |
Polyclonal | |
Rabbit | |
Phospho Specific | |
100 ul phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA. with 0.05% Sodium Azide | |
N-WASPneural WiskNWASP, WiskDKFZp779G0847, WiskMGC48327 | |
WASL | |
IgG | |
Affinity Purified | |
The antibody detects a 63 kDa protein corresponding to the molecular weight of phosphorylated WASP on SDS-PAGE immunoBlots of Jurkat cell lysate. In rat brain, A431, human endothelial and SKN-SH cells, this antibody detects a 65 kDa protein corresponding to phosphorylated N-WASP. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title