Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-WASP, p Tyr256 Antibody, Novus Biologicals™
SDP

Catalog No. NB6001169 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001169 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB6001169 Supplier Novus Biologicals Supplier No. NB6001169
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

N-WASP Polyclonal specifically detects N-WASP in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen N-WASP (p Tyr256)
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:10000, ELISA 1:20000
Formulation 0.1 ml PBS, 1.0 BSA mg/ml and 50% Glycerol with 0.05% Sodium Azide
Gene Alias N-WASPneural WiskNWASP, WiskDKFZp779G0847, WiskMGC48327
Gene Symbols WASL
Host Species Rabbit
Immunogen Phospho N WASP (Tyr 256) synthetic peptide (coupled to carrier protein) corresponding to amino acid residues around tyrosine 256 of human N WASP. The human WASP sequence has a similar peptide sequence surrounding tyrosine 291.
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 8976
Test Specificity N WASP [Tyr256]
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.