Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific Chemicals N4-Benzoylcytidine, 99%
Click to view available options
Quantity:
5 g
10 g
25 g
Description
N-Benzoylcytidine is a useful building block for oligoribonucleotide synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Benzoylcytidine is a useful building block for oligoribonucleotide synthesis.
Solubility
Soluble in 1N NaOH at 10mg/ml. Also soluble in methanol or DMSO
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
N-Benzoylcytidine is a useful building block for oligoribonucleotide synthesis.
Solubility
Soluble in 1N NaOH at 10mg/ml. Also soluble in methanol or DMSO
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Specifications
Specifications
Melting Point | 230°C to 234°C (decomposition) |
Color | White |
Quantity | 10 g |
Solubility Information | Soluble in 1N NaOH at 10mg/ml; Also soluble in methanol or DMSO |
Optical Rotation | +55° (c=0.1 in Methanol) |
Formula Weight | 347.32 |
Percent Purity | 99% |
Physical Form | Crystalline Powder |
Chemical Name or Material | N4-Benzoylcytidine |
Safety and Handling
TSCA | No |
Recommended Storage | Keep cold |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction