Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N4BP2L2 Antibody (OTI1C8), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB27286A488 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB27286A488 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB27286A488 Supplier Novus Biologicals Supplier No. NBP272866AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

N4BP2L2 Monoclonal specifically detects N4BP2L2 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen N4BP2L2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1C8
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CG005FLJ41089, CG016, FLJ36195, NEDD4 binding protein 2-like 2, NEDD4-binding protein 2-like 2, PFAAP5FLJ43077, Phosphonoformate immuno-associated protein 5,92M18.3,92M18.3 (novel protein), protein from BCRA2 region
Gene Symbols N4BP2L2
Host Species Mouse
Immunogen Full length recombinant protein of human N4BP2L2 (NP_055702) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 10443.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.