Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N6-methyladenosine (m6A) Antibody (17-3-4-1), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP250525FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250525FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250525FR Supplier Novus Biologicals Supplier No. NBP250525FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

N6-methyladenosine (m6A) Monoclonal specifically detects N6-methyladenosine (m6A) in Human, Mouse, Yeast samples. It is validated for Western Blot.

Specifications

Antigen N6-methyladenosine (m6A)
Applications Western Blot
Classification Monoclonal
Clone 17-3-4-1
Conjugate DyLight 680
Dilution Western Blot
Formulation PBS with 0.05% Sodium Azide
Host Species Mouse
Immunogen Hapten N6-methyladenosine-5'-mono-phosphate conjugated to BSA of all N6-methyladenosine
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Epigenetics, metabolism
Primary or Secondary Primary
Test Specificity This antibody recognizes N6-methyladenosine in both modified RNA AND DNA.
Target Species Human, Mouse, Yeast
Content And Storage Store at 4°C in the dark.
Isotype IgG1 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.