Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N6-methyladenosine (m6A) Antibody (17-3-4-1), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB309287 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309287 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309287 Supplier Novus Biologicals Supplier No. NBP250525MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

N6-methyladenosine (m6A) Monoclonal antibody specifically detects N6-methyladenosine (m6A) in Human, Mouse, Yeast samples. It is validated for Western Blot, Immunoprecipitation, Dot Blot

Specifications

Antigen N6-methyladenosine (m6A)
Applications Western Blot, Immunoprecipitation, Dot Blot
Classification Monoclonal
Clone 17-3-4-1
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunoprecipitation, Dot Blot
Formulation 50mM Sodium Borate
Gene Alias (-)-6-(Methylamino)purine riboside, N(6)-Methyladenosine, N-Methyladenosine
Host Species Mouse
Immunogen Hapten N6-methyladenosine-5'-mono-phosphate conjugated to BSA of all N6-methyladenosine
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epigenetics, metabolism
Primary or Secondary Primary
Target Species Human, Mouse, Yeast
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.