Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N6-Methyladenosine (m6A) Polyclonal Antibody, Active Motif™
SDP

Rabbit Polyclonal Antibody

$156.65 - $620.58

Specifications

Antigen N6-Methyladenosine (m6A)
Concentration Lot-Specific
Applications Dot Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-3450
SDP
Encompass
View Documents
Active Motif
61496
10 μg
Each for $156.65
Only null left
Add to Cart
 
50-227-7264
SDP
Encompass
View Documents
Active Motif
61995
50 μg
Each for $355.48
Only null left
Add to Cart
 
50-199-3449
SDP
Encompass
View Documents
Active Motif
61495
100 μg
Each for $620.58
Only null left
Add to Cart
 
Description

Description

N6-methyladenosine (m6A), or methylation of the N6 position of adenosine is a posttranscriptional modification of RNA. Due to a lack of analytical methods, N6-methyladenosine is poorly understood, but new evidence suggests that it is a very common base modification and important physiological regulator. N6-methyladenosine is markedly increased throughout brain development, and is enriched near stop codons, microRNA-binding sites and UTRs, which indicates a fundamental role in the regulation of gene expression. N6-methyladenosine is also highly conserved between human and mouse.
Specifications

Specifications

N6-Methyladenosine (m6A)
Dot Blot
Unconjugated
Rabbit
Human, Multi-species, Mouse, Rat
This antibody was raised against 6-Methyladenosine conjugated to kLH.
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lot-Specific
Polyclonal
Liquid
RUO
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
IgG
Protein A
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.