Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAA10 Antibody (3G3E9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261890
Description
NAA10 Monoclonal specifically detects NAA10 in Human, Mouse, Monkey samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
NAA10 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 | |
ARD1, ARD1 homolog A, N-acetyltransferase (S. cerevisiae), ARD1 homolog, N-acetyltransferase (S. cerevisiae), ARD1AARD1 homolog, N-acetyltransferase, EC 2.3.1.-, EC 2.3.1.88, MGC71248, N(alpha)-acetyltransferase 10, NatA catalytic subunit, N-acetyltransferase, N-acetyltransferase ARD1, human homolog of, N-alpha-acetyltransferase 10, NatA catalytic subunit, N-terminal acetyltransferase complex ARD1 subunit homolog A | |
Mouse | |
26.5 kDa | |
0.1 mL | |
Primary | |
Human, Mouse, Monkey | |
Purified |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
3G3E9 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
NAA10 | |
Purified recombinant fragment of human NAA10 (AA: 111-235) expressed in E. Coli. | |
Protein G purified | |
metabolism | |
8260 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction