Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAA10 Antibody (3G3E9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | NAA10 |
---|---|
Clone | 3G3E9 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61890
![]() |
Novus Biologicals
NBP261890 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
NAA10 Monoclonal specifically detects NAA10 in Human, Mouse, Monkey samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
NAA10 | |
1 mg/mL | |
Monoclonal | |
Purified | |
metabolism | |
ARD1, ARD1 homolog A, N-acetyltransferase (S. cerevisiae), ARD1 homolog, N-acetyltransferase (S. cerevisiae), ARD1AARD1 homolog, N-acetyltransferase, EC 2.3.1.-, EC 2.3.1.88, MGC71248, N(alpha)-acetyltransferase 10, NatA catalytic subunit, N-acetyltransferase, N-acetyltransferase ARD1, human homolog of, N-alpha-acetyltransferase 10, NatA catalytic subunit, N-terminal acetyltransferase complex ARD1 subunit homolog A | |
NAA10 | |
IgG1 | |
Protein G purified |
3G3E9 | |
Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 | |
Unconjugated | |
Mouse | |
Human, Mouse, Monkey | |
8260 | |
Purified recombinant fragment of human NAA10 (AA: 111-235) expressed in E. Coli. | |
Primary | |
26.5 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title