Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAALADase-like 1/NAALADL1 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB053555 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB053555 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB053555 Supplier Novus Biologicals Supplier No. NBP298362AF700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NAALADase-like 1/NAALADL1 Polyclonal specifically detects NAALADase-like 1/NAALADL1 in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen NAALADase-like 1/NAALADL1
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias I100, NAALADASEL, NAALADL1 N-acetylated alpha-linked acidic dipeptidase-like 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant NAALADase-like 1/NAALADL1 (Pro29-Leu745).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10004
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.