Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAB1 Monoclonal Antibody (OTI1B1), TrueMAB™, OriGene
SDP

Catalog No. 501679484
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-9484 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-9484 Supplier OriGene Supplier No. TA802909
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Acts as a transcriptional repressor for zinc finger transcription factors EGR1 and EGR2.

Specifications

Antigen NAB1
Applications Western Blot
Classification Monoclonal
Clone OTI1B1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene NAB1
Gene Accession No. Q13506
Gene Alias NGFI-A binding protein 1
Gene Symbols NAB1
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 237-487 of human NAB1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4664
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.