Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nab2 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB324588 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB324588 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB324588 Supplier Novus Biologicals Supplier No. NBP299585MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Nab2 Polyclonal antibody specifically detects Nab2 in Human samples. It is validated for Western Blot

Specifications

Antigen Nab2
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias EGR-1-binding protein 2, MADEREGR1 binding protein 2, Melanoma-associated delayed early response protein, MGC75085, NGFI-A binding protein 2 (EGR1 binding protein 2), NGFI-A-binding protein 2, Protein MADER
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Nab2.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 4665
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.