Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAC1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10058964 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10058964 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10058964 Supplier Novus Biologicals Supplier No. NB10058964
Add to Cart
Add to Cart

Goat Polyclonal Antibody

NAC1 Polyclonal specifically detects NAC1 in Human samples. It is validated for Peptide ELISA, Western Blot.

Specifications

Antigen NAC1
Applications Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1.5 ug/ml, Peptide ELISA Detection limit 1:2000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_443108.1
Gene Alias BEND8, BTB (POZ) domain containing 14B, BTB/POZ domain-containing protein 14B, BTBD14B, NAC1BEN domain containing 8, NAC-1FLJ37383, nucleus accumbens associated 1, BEN and BTB (POZ) domain containing, nucleus accumbens-associated protein 1, transcriptional repressor NAC1
Gene Symbols NACC1
Host Species Goat
Immunogen Peptide with sequence KTEQQESDSVQC corresponding to internal region according to NP_443108.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112939
Test Specificity Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog and Cow.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.