Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAC1 Antibody (PCRP-NACC1-1A8), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB390238 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB390238 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB390238 Supplier Novus Biologicals Supplier No. NBP308833AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NAC1 Monoclonal antibody specifically detects NAC1 in Human samples. It is validated for Flow Cytometry, Protein Array

Specifications

Antigen NAC1
Applications Flow Cytometry, Protein Array
Classification Monoclonal
Clone PCRP-NACC1-1A8
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias BEND8, BTB (POZ) domain containing 14B, BTB/POZ domain-containing protein 14B, BTBD14B, NAC1BEN domain containing 8, NAC-1FLJ37383, nucleus accumbens associated 1, BEN and BTB (POZ) domain containing, nucleus accumbens-associated protein 1, transcriptional repressor NAC1
Host Species Mouse
Immunogen Recombinant full-length human NAC1 protein (Uniprot: Q96RE7)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Stem Cell Markers, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 112939
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.