Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Nafamostat mesylate
GREENER_CHOICESDP

Catalog No. 308150 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-815-0 50 mg
30-811-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 30-815-0 Supplier R&D Systems™ Supplier No. 3081/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Serine protease inhibitor, inhibits tryptase

Nafamostat mesylate is a broad spectrum serine protease inhibitor. Displays selectivity for human tryptase when used at relatively low concentrations. Reduces eosinophil infiltration, mast cell activation and airway responsiveness in a murine model of asthma. Inhibits SARS-CoV-2 infection in vitro (EC50 = 22.50 μM).

R&D Systems products are for biomedical research use only. They are not intended for human or veterinary use.

Chemical Identifiers

CAS 82956-11-4
Molecular Formula C21H25N5O8S2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 539.59
Purity >99%
Molecular Weight (g/mol) 539.58
Chemical Name or Material Nafamostat mesylate
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.