Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAIP Antibody (13E6), Novus Biologicals™
SDP

Catalog No. NB298736 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298736 50 μL
NB298737 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298736 Supplier Novus Biologicals Supplier No. NBP32612050UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

NAIP Monoclonal antibody specifically detects NAIP in Human samples. It is validated for ELISA, Immunohistochemistry

Specifications

Antigen NAIP
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 13E6
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias baculoviral IAP repeat-containing 1, BIRC1baculoviral IAP repeat-containing protein 1, FLJ18088, FLJ42520, FLJ58811, Neuronal apoptosis inhibitory protein, NLR family, apoptosis inhibitory protein, NLR family, BIR domain containing 1, NLRB1, nucleotide-binding oligomerization domain, leucine rich repeat and BIR domaincontaining 1, psi neuronal apoptosis inhibitory protein, psiNAIP
Host Species Rabbit
Immunogen A synthesized peptide derived from Human NAIP [UniProt Q13075]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 4671
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.