Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naip1/2/5 Antibody (Naipa-1), Biotin, Novus Biologicals™
SDP

Catalog No. NB173049 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB173049 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB173049 Supplier Novus Biologicals Supplier No. NBP280079B
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Naip1/2/5 Monoclonal antibody specifically detects Naip1/2/5 in Mouse samples. It is validated for Western Blot

Specifications

Antigen Naip1/2/5
Applications Western Blot
Classification Monoclonal
Clone Naipa-1
Conjugate Biotin
Formulation PBS
Gene Alias baculoviral IAP repeat-containing 1a, baculoviral IAP repeat-containing 1e, baculoviral IAP repeat-containing protein 1a, baculoviral IAP repeat-containing protein 1b, baculoviral IAP repeat-containing protein 1e, Birc1a, Birc1b, Birc1e, D13Lsd1, Legionella, susceptiblility 1, Lgn1, Naip, Naip-rs1, Naip-rs3, Naip-rs6, neuronal apoptosis inhibitory protein 1, neuronal apoptosis inhibitory protein 5, neuronal apoptosis inhibitory protein, related sequence 3, neuronal apoptosis inhibitory protein, related sequence 6
Host Species Mouse
Immunogen Recombinant mouse NAIP5 (BIR2 domain/aa 159-227).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 17940
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.