Learn More
Thermo Scientific™ Nalgene™ Autoclavable Graduated Buckets
Description
Contain a variety of substances, including chemical and corrosive liquids, in sturdy and convenient Thermo Scientific™ Nalgene™ Autoclavable Graduated Buckets, which are a lab necessity. These general-purpose, polypropylene laboratory buckets are fully autoclavable and provide excellent chemical resistance.
- White, polypropylene pails offer sturdy construction for long-lasting use in the lab.
- Nylon-coated, wire bail handle provides a comfortable and sturdy grip.
- Molded-in graduated in pints and liters are easy to read.
Specifications
Specifications
| Capacity (Metric) | 10.4 L |
| Graduations | Molded |
| Material | Polypropylene |
| Autoclavable | Autoclavable |
| Capacity (English) | 11 qt. |
| Description | Bucket Graduated PP 10.4 L |
| Includes | Handle |
| Quantity | 1 Ea. |