Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Centrifuge Tubes made with Teflon™ fluoropolymer and Closure made with Tefzel™
GSA_VAGREENER_CHOICE

Catalog No. 0556216c
Click to view available options
Capacity (Metric):
10 mL
30 mL
50 mL
Diameter (Metric):
16 mm
25.7 mm
28.8 mm
Note: Must be filled to 100% of total capacity for centrifugation
Note: Must be filled to 100% of total capacity for centrifugation

Conduct high-speed centrifugation of aggressive chemicals with these tubes, suited for chloroform and phenol extractions in refrigerated machines up to 50,000 x g.

Conduct high-speed centrifugation of aggressive chemicals with Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Centrifuge Tubes made with Teflon™ fluoropolymer and Closure made with Tefzel™, specifically suited for chloroform and phenol extractions.

Features:

  • Teflon™ FEP, Tefzel™ ETFE screw closure
  • Withstand temperatures from -100° to +150°C (-148° to +302°F)
  • Leakproof† Tefzel ETFE closures prevent material loss under ordinary use for most spins <10,000 × g
  • Sealing caps available for 30mL and 50mL size to assure leakproof service especially at high speeds >10,000 × g or when spinning hazardous materials
  • Autoclavable/biohazard/leakproof†/PTFE FEP

Ordering Information:

Optional sealing caps available: DS3131-0020 (for 30mL tube) and DS3131-0024 (for 50mL tube). Cat. No. 3114-0050 has a brim capacity of only 42mL but is designed to fit standard rotor.

Includes:

Tefzel ETFE screw caps. Additional sealing cap assembly available separately (DS3131-series).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Closure Material ETFE
Capacity (Metric) 10 mL
Material FEP
No. per Case 10
No. per Pack 2
Sterility Non-sterile
Autoclavable Autoclavable
Shape Round
Bottom Shape Round
Closure Type Screw Cap
Writing Area No
Diameter (Metric) 16 mm
Graduated No
Product Type Centrifuge Tube
Max. RCF 10,000 x g
Show More Show Less

Teflon™ is a trademark of The Chemours Company FC, LLC used under license by Nalge Nunc International Corporation.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes.

Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.