Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Polycarbonate Centrifuge Tubes w/Sealing Cap
GSA_VAGREENER_CHOICE

Catalog No. p-4526055
Click to view available options
Capacity (Metric):
10 mL
16 mL
30 mL
50 mL
Length (Metric):
83.8 mm
102.6 mm
107.2 mm
114.8 mm
Diameter (Metric):
16.1 mm
18.2 mm
25.8 mm
28.8 mm
Autoclaving repeatedly will shorten the life of polycarbonate tubes. Tubes must be filled to at least 80% capacity for proper performance.
Autoclaving repeatedly will shorten the life of polycarbonate tubes. Tubes must be filled to at least 80% capacity for proper performance.

View contents easily using these strong, transparent polycarbonate centrifuge tubes, available with sealing caps for use up to 50,000 x g.

Easily view contents using strong, transparent Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Polycarbonate Centrifuge Tubes, equipped with sealing caps for leakproof† performance up to 50,000 x g.

Features:
  • Withstand temperatures from -135° to +135°C (-211° to +275°F)
  • Round bottom to fit high speed rotors
  • Can be used up to 50,000 x g in refrigerated or non-refrigerated centrifuges
  • Come with PP sealing caps with silicone gaskets for extra leak protection when spinning >10,000 x g or spinning hazardous materials
  • Autoclavable; transparent

Ordering Information:

Sealing caps also sold separately for 30 and 50mL sizes.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Closure Material Polypropylene, red rubber
Capacity (Metric) 30 mL
Material Polycarbonate
No. per Case 50
No. per Pack 10
Sterility Non-sterile
Autoclavable Autoclavable
Shape Round
Length (Metric) 102.6 mm
Bottom Shape Round
Closure Type Screw Cap
Writing Area No
Diameter (Metric) 25.8 mm
Graduated No
Product Type Centrifuge Tube
Max. RCF 50,000 x g
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.