Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Nalgene™ PPCO Conical-Bottom Centrifuge Tubes

Click to view available options
Capacity (Metric):
15 mL
50 mL
Length (Metric):
118.6 mm
133.1 mm
Diameter (Metric):
16.8 mm
28.7 mm
Description
Simplify measurements using Thermo Scientific™ Nalgene™ PPCO Conical-Bottom Centrifuge Tubes, which feature molded-in graduations. Translucent polypropylene copolymer (PPCO) has excellent chemical resistance.
Features:
- Translucent Polypropylene Copolymer (PPCO) is compatible with a wide range of lab reagents
- Designed for low-speed centrifugation up to 6000 × g in refrigerated or non-refrigerated centrifuges (tubes must be filled at least 80% for proper performance)
- Conical bottoms concentrate pellet in a small area for easy isolation and retrieval
- Molded-in graduations last the life of the tube
- Last longer than polycarbonate tubes under conditions of repeated autoclaving
- Tube are both autoclavable and graduated
Ordering Information:
Supplied without closure; friction fit closures available. For 15mL use DS3111-00017; for 50mL use DS3111-0029.

Specifications
Specifications
Capacity (Metric) | 15 mL |
Material | Polypropylene Copolymer |
No. per Case | 100 |
No. per Pack | 10 |
Sterility | Non-sterile |
Autoclavable | Autoclavable |
Shape | Conical |
Length (Metric) | 118.6 mm |
Bottom Shape | Conical |
Writing Area | No |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction