Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NALP12 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB358522 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB358522 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB358522 Supplier Novus Biologicals Supplier No. NBP176293PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NALP12 Polyclonal antibody specifically detects NALP12 in Human, Mouse, Rat, Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen NALP12
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias FCAS2, Monarch-1, NACHT, leucine rich repeat and PYD containing 12, NACHT, LRR and PYD domains-containing protein 12, NALP12LRR and PYD containing protein 12, NLR family, pyrin domain containing 12, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 12, PYPAF7Monarch1, PYRIN-containing APAF1-like protein 7, Regulated by nitric oxide, RNO2
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human NALP12. The immunogen is located within amino acids 110 - 160 of NALP12. Amino Acid Squence: TPRKDPQETYRDYVRRKF
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 91662
Target Species Human, Mouse, Rat, Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.