Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NALP6 Antibody (920631), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB345854 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345854 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345854 Supplier Novus Biologicals Supplier No. FAB9145MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NALP6 Monoclonal specifically detects NALP6 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen NALP6
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 920631
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Angiotensin II/Vasopressin Receptor, AVR, CLR11.4, NACHT, Leucine Rich Repeat And PYD Containing 6, NACHT, LRR And PYD Containing Protein 6, NACHT, LRR And PYD Domains-Containing Protein 6, NAVR, NAVR/AVR, NLR Family, Pyrin Domain Containing 6, NLRP6, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And Pyrin Domain Containing 6, PAN3, PYPAF5, PYRIN-Containing APAF1-Like Protein 5
Host Species Mouse
Immunogen E.coli-derived recombinant human NLRP6/NALP6, Lys35-Arg193, Accession # P59044
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Inflammation
Primary or Secondary Primary
Gene ID (Entrez) 171389
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.