Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NALP6 Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB191858 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB191858 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB191858 Supplier Novus Biologicals Supplier No. NBP281856JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NALP6 Polyclonal antibody specifically detects NALP6 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen NALP6
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias Angiotensin II/Vasopressin Receptor, AVR, CLR11.4, NACHT, Leucine Rich Repeat And PYD Containing 6, NACHT, LRR And PYD Containing Protein 6, NACHT, LRR And PYD Domains-Containing Protein 6, NAVR, NAVR/AVR, NLR Family, Pyrin Domain Containing 6, NLRP6, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And Pyrin Domain Containing 6, PAN3, PYPAF5, PYRIN-Containing APAF1-Like Protein 5
Host Species Rabbit
Immunogen NALP6 antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human NALP6. The immunogen is located within amino acids 150 - 200 of NALP6. Amino Acid Squence: AEEPEPGRARRSDTHRT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Inflammation
Primary or Secondary Primary
Gene ID (Entrez) 171389
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.