Learn More
Promega Nano-Glo™ Dual-Luciferase™ Reporter Assay/pNL1.1.PGK Bundle
Catalog No. prn1531 Shop All Promega Corporation Products
Click to view available options
Quantity:
1 Ea.
The Nano-Glo™ Dual-Luciferase Reporter (NanoDLR) Assay System is a homogeneous reagent system that allows you to sequentially detect the activities of firefly (Photinus pyralis) luciferase and NanoLuc luciferase (Nluc) from a single sample.
  • Ultra-Sensitive Detection of Firefly and NanoLuc(R) Luciferase Activities in a Single Sample
  • Greater sensitivity at low expression levels
  • Choose either firefly or NanoLuc(R) luciferase as primary reporters
  • Improved reagent stability brings more flexibility to assay design
Specifications
Quantity 1 Ea.
Content And Storage Varies Among Components
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.