Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Nano-Glo In-Gel Detection System, 100ml

Catalog No. prn3020 Shop All Promega Corporation Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRN3020 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PRN3020 Supplier Promega Supplier No. N3020
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

The Nano-Glo In-Gel Detection System directly detects the luminescence of NanoLuc fusion proteins in polyacrylamide gels after protein separation by PAGE under native or denaturing SDS conditions (native PAGE or SDS-PAGE).

  • Directly Detect NanoLuc(R) Fusion Proteins in Polyacrylamide Gels
  • Simply incubate native or SDS denaturing gels with reagent and image
  • No transfer to membranes required for detection
  • Eliminates the need for blocking or antibodies

Specifications

Quantity 100 mL
Storage Requirements -30°C to -10°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.