Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nanog Antibody (1E6C4), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP147427A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP147427A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP147427A30 Supplier Novus Biologicals Supplier No. NBP147427AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nanog Monoclonal specifically detects Nanog in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Nanog
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 1E6C4
Conjugate Alexa Fluor 350
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias FLJ12581, FLJ40451, hNanog, homeobox protein NANOG, Homeobox transcription factor Nanog, homeobox transcription factor Nanog-delta 48, Nanog homeobox
Gene Symbols NANOG
Host Species Mouse
Immunogen Purified recombinant fragment of human Nanog (amino acids 20-166) expressed in E. coli. [UniProtNo. Q9H9S0]
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Apoptosis, Autophagy, Cancer, Cellular Markers, Embryonic Stem Cell Markers, Macroautophagy, mTOR Pathway, Neurodegeneration, Protein Turnover, Regulatory Immunology, Signal Transduction, Stem Cell Markers, Stem Cell Signaling Pathway, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 79923.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.