Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYH Antibody, Novus Biologicals™
SDP

Catalog No. NB6001032SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001032SS 0.025 mg
NB6001032 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB6001032SS Supplier Novus Biologicals Supplier No. NB6001032SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 7 publications

MYH Polyclonal specifically detects MYH in Human, Mouse, Rat, Bacteria samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen MYH
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml, Immunohistochemistry 1:10- 1:2000. Use reported in scientific literature, Immunocytochemistry/Immunofluorescence 18 ug/ml. Use reported in scientific literature
Formulation PBS containing 0.05% BSA with 0.02% Sodium Azide
Gene Accession No. NP_001121897
Gene Alias CYP2C, EC 3.2.2, EC 3.2.2.-, hMYH, MGC4416, mutY (E. coli) homolog, MutY homolog, mutY homolog (E. coli), Muyth, MYHA/G-specific adenine DNA glycosylase
Gene Symbols MUTYH
Host Species Rabbit
Immunogen This antibody was generated by immunizing rabbits with a synthetic peptide corresponding to amino acids 531-546 of human MYH.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Base Excision Repair, Cancer, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 4595
Test Specificity Cross-reacts with Human, Mouse, Rat and Bacteria. Not yet tested in other species.
Target Species Human, Mouse, Rat, Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.