Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nanog Antibody, Novus Biologicals™
SDP

Catalog No. NBP224941SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224941SS 0.025 mg
NBP224941 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224941SS Supplier Novus Biologicals Supplier No. NBP224941SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

Nanog Polyclonal specifically detects Nanog in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Nanog
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2 ug/ml, Flow Cytometry reported in scientific literature (PMID 26283864), Immunocytochemistry/Immunofluorescence 1:10
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q9H9S0
Gene Alias FLJ12581, FLJ40451, hNanog, homeobox protein NANOG, Homeobox transcription factor Nanog, homeobox transcription factor Nanog-delta 48, Nanog homeobox
Gene Symbols NANOG
Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human NANOG was used as the immunogen.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cellular Markers, Embryonic Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 79923
Test Specificity This antibody is reactive to Chimpanzee, Goat, Human, Mouse, Rhesus Monkey.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.