Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nanos2 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP176370 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176370 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176370 Supplier Novus Biologicals Supplier No. NBP176370
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Nanos2 Polyclonal specifically detects Nanos2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Nanos2
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-4 μg/mL, ELISA 1:100-1:2000
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. P60321
Gene Alias nanos homolog 2, nanos homolog 2 (Drosophila), NOS2NOS-2
Gene Symbols NANOS2
Host Species Rabbit
Immunogen Antibody was raised against a 13 amino acid synthetic peptide near the carboxy terminus of the human Nanos2. The immunogen is located within the last 50 amino acids of Nanos2. Amino Acid Squence: QSLYRRSGRNSAGRR
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339345
Test Specificity This Nanos2 antibody will not cross-react with either Nanos 1 or Nanos3.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.