Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NANOS3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | NANOS3 |
|---|---|
| Concentration | 0.33 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-3547
![]()
|
Proteintech Group Inc
216791AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
os is a zinc-finger containing, RNA-binding protein that has been implicated in germ cell development in both invertebrates and vertebrates. In Drosophila, os represses apoptosis during development to ensure proper germ-line development. Unlike os1 whose expression in mice is dispensable, the os2 and os3 proteins are required for germ cell development. os2-null primordial germ cells (PGCs) die only in the male gonads and show no defects in females, while os3-null PGCs are lost during the migration stage regardless of sex. os2 and os3 have distinct expression patterns during embryo development, suggesting that these two proteins do not have redundant functions. However, expression of os2 can at least partially replace os3 function in a os3-null background. os3 expression can not rescue os2-null defects.Specifications
| NANOS3 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P60323, P60324 | |
| 244551, 288909, 342977 | |
| NANOS3 Fusion Protein Ag16014 | |
| Primary | |
| -20°C | |
| NANOS3 |
| 0.33 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| os homolog 3, os homolog 3 (Drosophila), NANOS1L, NANOS3, NOS 3, NOS3 | |
| os3 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title