Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NANP Antibody (OTI4D11), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP272870A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP272870A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP272870A30 Supplier Novus Biologicals Supplier No. NBP272870AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NANP Monoclonal specifically detects NANP in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen NANP
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI4D11
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias C20orf147, chromosome 20 open reading frame 147, dJ694B14.3, EC 3.1.3.29, haloacid dehalogenase-like hydrolase domain containing 4, Haloacid dehalogenase-like hydrolase domain-containing protein 4, HDHD4Neu5Ac-9-Pase, MGC26833, N-acetylneuraminic acid phosphatase, N-acylneuraminate-9-phosphatase
Gene Symbols NANP
Host Species Mouse
Immunogen Full length recombinant protein of human NANP (NP_689880) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 140838
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.