Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAP1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017373 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017373 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017373 Supplier Novus Biologicals Supplier No. NBP282062
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NAP1 Polyclonal specifically detects NAP1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen NAP1
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias CG5330, Dmel\CG5330, dNap, dNAP1, dNAP-1, MAP3K7IP3, MGC45404, mitogen-activated protein kinase kinase kinase 7 interacting protein 3, Mitogen-activated protein kinase kinase kinase 7-interacting protein 3, NAP1, NAP-1, NF-kappa-B-activating protein 1, NFkB activating protein 1, p56/dCAF-4, TAB-3, TAK1-binding protein 3, TGF-beta activated kinase 1/MAP3K7 binding protein 3, TGF-beta-activated kinase 1 and MAP3K7-binding protein 3, TGF-beta-activated kinase 1-binding protein 3
Gene Symbols TAB3
Host Species Rabbit
Immunogen NAP1 antibody was raised against a 13 amino acid synthetic peptide near the amino terminus of human NAP1. The immunogen is located within amino acids 40 - 90 of NAP1. Amino Acid Squence: YHSPDDNRMNRNR
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 257397
Test Specificity NAP1 antibody is human specific. NAP1 antibody is predicted not to cross-react with other TAB proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.