Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAP1L3 Antibody (OTI3G3), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. N272875J549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272875J549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272875J549 Supplier Novus Biologicals Supplier No. NBP272875JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NAP1L3 Monoclonal specifically detects NAP1L3 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen NAP1L3
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3G3
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias BNAP, MB20, MGC26312, NPL3, nucleosome assembly protein 1-like 3
Gene Symbols NAP1L3
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-315 of human NAP1L3 (NP_004529) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 4675.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.