Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAPA Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB087483 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB087483 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB087483 Supplier Novus Biologicals Supplier No. NBP300366B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NAPA Polyclonal specifically detects NAPA in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen NAPA
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias alpha-SNAP, alpha-soluble NSF attachment protein, N-ethylmaleimide-sensitive factor attachment protein alpha, N-ethylmaleimide-sensitive factor attachment protein, alpha, S, SNAPA, SNAP-alpha
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant NAPA (NP_003818.2; Met1-Arg295).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8775
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.