Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Napabucasin

Click to view available options
Quantity:
10 mg
50 mg
Description
Napabucasin is a STAT3 inhibitor. Blocks spherogenesis of cancer stem cells (CSC), but not hematopoietic stem cells. Inhibits PaCa-2 xenograft tumor growth in mice, even after cessation of treatment, and decreases population of CSCs in PaCa-2 tumors from treated mice. Also blocks spleen and liver metastases of colon cancer cells in a mouse model.
Specifications
Specifications
| Chemical Name or Material | 2-Acetylnaphtho[2,3-b]furan-4,9-dione |
| CAS | 83280-65-3 |
| Quantity | 10 mg |
| Target | STAT Inhibitors |
| Molecular Formula | C14H8O4 |
| Formula Weight | 240.21 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction