Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAPE-PLD Antibody, Novus Biologicals™
SDP

Catalog No. NB11080070T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11080070T 0.025 mL
NB11080070 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11080070T Supplier Novus Biologicals Supplier No. NB11080070SS

Rabbit Polyclonal Antibody has been used in 8 publications

NAPE-PLD Polyclonal specifically detects NAPE-PLD in Human, Mouse, Rat, Fungi samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen NAPE-PLD
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:2500-1:5000, Immunocytochemistry/Immunofluorescence 1:1000-1:5000
Formulation Whole antisera with 0.1% Sodium Azide
Gene Accession No. Q8BH82
Gene Alias C7orf18, DKFZp781D1098, EC 3.1.4, EC 3.1.4.-, N-acyl phosphatidylethanolamine phospholipase DNAPE-PLDFMP30, N-acyl-phosphatidylethanolamine-hydrolyzing phospholipase D, NAPE-hydrolyzing phospholipase D
Gene Symbols NAPEPLD
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of mouse NAPE-PLD (within residues 1-100). [Swiss-Prot# Q8BH82]
Molecular Weight of Antigen 50 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 222236
Test Specificity Human, Mouse and Rat. Fungi reactivity reported in scientific literature (PMID: 25433633).
Target Species Human, Mouse, Rat, Fungi
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.