Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAPE-PLD Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 8 publications
$206.00
Specifications
Antigen | NAPE-PLD |
---|---|
Dilution | Western Blot 1:2500-1:5000, Immunocytochemistry/Immunofluorescence 1:1000-1:5000 |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11080070T
![]() |
Novus Biologicals
NB11080070SS |
0.025 mL |
Each for $206.00
|
|
|||||
NB11080070
![]() |
Novus Biologicals
NB11080070 |
0.1 mL | N/A | N/A | N/A | ||||
Description
NAPE-PLD Polyclonal specifically detects NAPE-PLD in Human, Mouse, Rat, Fungi samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
NAPE-PLD | |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism, Signal Transduction | |
Whole antisera with 0.1% Sodium Azide | |
C7orf18, DKFZp781D1098, EC 3.1.4, EC 3.1.4.-, N-acyl phosphatidylethanolamine phospholipase DNAPE-PLDFMP30, N-acyl-phosphatidylethanolamine-hydrolyzing phospholipase D, NAPE-hydrolyzing phospholipase D | |
NAPEPLD | |
IgG | |
Unpurified | |
Human, Mouse and Rat. Fungi reactivity reported in scientific literature (PMID: 25433633). |
Western Blot 1:2500-1:5000, Immunocytochemistry/Immunofluorescence 1:1000-1:5000 | |
Polyclonal | |
Antisera | |
RUO | |
Human, Mouse, Rat, Fungi | |
Q8BH82 | |
222236 | |
Synthetic peptide made to an internal portion of mouse NAPE-PLD (within residues 1-100). [Swiss-Prot# Q8BH82] | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. | |
50 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title