Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naphthalene-1,4-dicarboxylic acid, 98+%

Catalog No. p-7048113
Click to view available options
Quantity:
25 g
100 g

Catalog No. AAL2024914

Supplier: Thermo Scientific Chemicals L2024914

Only null left
Add to Cart

CAS: 605-70-9 | C12H8O4 | 216.19 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store away from water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 605-70-9
Molecular Formula C12H8O4
Molecular Weight (g/mol) 216.19
MDL Number MFCD00014312
InChI Key ABMFBCRYHDZLRD-UHFFFAOYSA-N
Synonym 1,4-naphthalenedicarboxylic acid, 1,4-naphthalenedicarboxylicacid, 1,4-naphthalicacid, 1,4-naphthalic acid, pubchem21317, acmc-1b0c7, ksc357a6b, naphthalene-1,4-dicarboxylicacid, naphthalene-1,4-dicarboxlic acid, naphthaline-1,4-dicarboxylic acid
PubChem CID 69065
IUPAC Name naphthalene-1,4-dicarboxylic acid
SMILES OC(=O)C1=C2C=CC=CC2=C(C=C1)C(O)=O

Specifications

Melting Point 321°C to 325°C
Quantity 25 g
Beilstein 2051255
Solubility Information Insoluble in water.
Formula Weight 216.19
Percent Purity ≥98%
Chemical Name or Material Naphthalene-1,4-dicarboxylic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 210-094-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.