Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NARC-1 (PCSK9) Mouse anti-Human, Unconjugated, Clone: 13D3, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABS216025UG
Description
NARC-1 (PCSK9) Monoclonal antibody specifically detects NARC-1 (PCSK9) in Human samples. It is validated for ELISA, Western Blotting.
Specifications
NARC-1 (PCSK9) | |
Monoclonal | |
Unconjugated | |
Proprotein convertase subtilisin/kexin type 9;Neural apoptosis-regulated convertase 1;Proprotein convertase 9;PC9;Subtilisin/kexin-like protease PC9. | |
Mouse | |
∼75 and 59 kDa observed (proprotein and cleaved form); 74.29 kDa calculated. Uncharacterized bands may be observed in some lysate(s). | |
25 μg | |
Primary | |
Human | |
Purified |
ELISA, Western Blot | |
13D3 | |
NP_777596 | |
PCSK9, NARC1, PSEC0052 | |
Full-length purified human Proprotein convertase 9 (PCSK9). | |
Protein G Purified | |
Signaling | |
Clone 13D3 is a mouse monoclonal antibody that specifically detects Proprotein convertase 9 (NARC-1) and its cleaved form in human cells. | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction