Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NARC-1 (PCSK9) Mouse anti-Human, Unconjugated, Clone: 13D3, MilliporeSigma™
Mouse Monoclonal Antibody
$189.29 - $477.80
Specifications
Antigen | NARC-1 (PCSK9) |
---|---|
Clone | 13D3 |
Applications | ELISA, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABS21602MI
|
MilliporeSigma
MABS216025UG |
25 μg |
Each for $189.29
|
|
|||||
MABS21601MI
|
MilliporeSigma
MABS2160100UG |
100 μg |
Each for $477.80
|
|
|||||
Description
NARC-1 (PCSK9) Monoclonal antibody specifically detects NARC-1 (PCSK9) in Human samples. It is validated for ELISA, Western Blotting.Specifications
NARC-1 (PCSK9) | |
ELISA, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Proprotein convertase subtilisin/kexin type 9;Neural apoptosis-regulated convertase 1;Proprotein convertase 9;PC9;Subtilisin/kexin-like protease PC9. | |
Full-length purified human Proprotein convertase 9 (PCSK9). | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
∼75 and 59 kDa observed (proprotein and cleaved form); 74.29 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
13D3 | |
Monoclonal | |
Purified | |
Signaling | |
NP_777596 | |
PCSK9, NARC1, PSEC0052 | |
IgG1 κ | |
Protein G Purified | |
Clone 13D3 is a mouse monoclonal antibody that specifically detects Proprotein convertase 9 (NARC-1) and its cleaved form in human cells. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title