Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NARFL Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB352803 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB352803 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB352803 Supplier Novus Biologicals Supplier No. NBP306258PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NARFL Polyclonal antibody specifically detects NARFL in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen NARFL
Applications Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias cytosolic Fe-S cluster assembly factor NARFL, FLJ21988, HPRN, IOP1Protein related to Narf, Iron-only hydrogenase-like protein 1, LET1 like/JFP15, nuclear prelamin A recognition factor-like, Nuclear prelamin A recognition factor-like protein, PRNLET1L
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human NARFL fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64428
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.