Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NARG1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP121387 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP121387 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP121387 Supplier Novus Biologicals Supplier No. NBP121387
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NARG1 Polyclonal specifically detects NARG1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen NARG1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:50 to 1:250, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:10-1:500
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. NP_476516.1
Gene Alias FLJ13340, GA19, Gastric cancer antigen Ga19, N(alpha)-acetyltransferase 15, NatA auxiliary subunit, NARG1, NATHGa19, NMDA receptor-regulated protein 1, N-terminal acetyltransferase, Protein tubedown-1, Tbdn100, TBDN100NatA auxiliary subunit, transcriptional coactivator tubedown-100, tubedown-1
Gene Symbols NAA15
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 816 and 866 of human NMDA receptor regulated 1 using the numbering given in entry NP_476516.1 (GeneID 80155).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis
Primary or Secondary Primary
Gene ID (Entrez) 80155
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, Crab-eating macaque and Northern white-cheeked gibbon.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.